missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Antibody (TB01), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62203
This item is not returnable.
View return policy
Description
CD57 Monoclonal antibody specifically detects CD57 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow (Cell Surface)
Specifications
| CD57 | |
| Monoclonal | |
| Unconjugated | |
| TBS | |
| Mouse | |
| Chromatography | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| TB01 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1-4 μg/ml, Immunohistochemistry 5-10 μg/ml, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Cell Surface) | |
| HNK1, HNK-1, Human Natural Killer-1, L2, LEU7, LEU-7 | |
| A pool of neuroblastoma cell lines | |
| 0.1 mg | |
| Cancer, Cell Biology, Immunology | |
| 27087 | |
| Store at 4°C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction