missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD57 Monoclonal Antibody (6-13-19-1)

Product Code. 11540762
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11540762 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11540762 Supplier Invitrogen™ Supplier No. MA135253

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The antibody reacts with a 110 kD antigen present on the peripheral blood lymphocyte subset (primarily large granular lymphocytes) identified by the HNK-1 monoclonal antibody. It recognizes part of the Natural Killer cell population. CD57 is furthermore present on cells in neuroectodermal tissue.

CD57, also known as HNK-1 and Leu-7, is an oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitin sulphate proteoglycans. Its function is poorly understood. CD57 is expressed on a subset of natural killer (NK) cells and NK T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD57
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 6-13-19-1
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene B3GAT1
Gene Accession No. Q9P2W7
Gene Alias HNK-1; Leu-7
Gene Symbols B3GAT1
Host Species Mouse
Immunogen JURKAT T-cell line.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.