Learn More
CD58/LFA-3 Antibody (13G5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26677-50ul
 View more versions of this product
Product Code. 30043917
  • € 343.00 / 50µL
Description
CD58/LFA-3 Monoclonal antibody specifically detects CD58/LFA-3 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD58/LFA-3
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA
13G5
Western Blot 1:500-1:2000, ELISA
Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3
A synthesized peptide derived from Human CD58/LFA-3 [UniProt P19256]
50 μL
Cell Biology, Cellular Markers, Immunology
965
Store at -20 to -70C. Avoid freeze-thaw cycles.