missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD58/LFA-3 Antibody (248310), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1689PECY55
This item is not returnable.
View return policy
Description
CD58/LFA-3 Monoclonal antibody specifically detects CD58/LFA-3 in Human samples. It is validated for Flow Cytometry
Specifications
| CD58/LFA-3 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 248310 | |
| Flow Cytometry | |
| Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 | |
| Mouse myeloma cell line NS0-derived recombinant human CD58/LFA-3, Phe29-Arg215, Accession # P19256 | |
| 0.5 mL | |
| Cell Biology, Cellular Markers, Immunology | |
| 965 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction