missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD58 Mouse anti-Human, BV480, Clone: 1C3 (also known as AICD58.6), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746658
This item is not returnable.
View return policy
Description
CD58 Monoclonal antibody specifically detects CD58 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD58 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1C3 (also known as AICD58.6) | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction