missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD58 Recombinant Rabbit Monoclonal Antibody (083), PerCP

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA546750

Product Code. 17850926

  • € 431.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD58 (Met1-Arg215) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

CD58 is a member of the immunoglobulin superfamily and exists both as a transmembrane protein and a GPI-linked extracellular protein, with cells capable of expressing both forms concurrently. Additionally, soluble CD58 has been detected in serum, urine, and synovial fluid, particularly in patients with rheumatoid arthritis. CD58 is widely expressed on non-hematopoietic and most hematopoietic cells. CD58 serves as a ligand for the CD2 protein on T lymphocytes, playing a crucial role in adhesion and activation of T cells. The interaction between CD58 and CD2 initiates a signaling pathway that includes phosphorylation and leads to cytokine release. This interaction is important for T cell activation, cytokine production, cytotoxicity, and adhesion to various cell types. The protein is localized to the plasma membrane, and alternatively spliced transcript variants have been described, indicating diverse functional roles. CD58's involvement in immune responses highlights its significance in both normal physiological processes and disease states.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD58
Recombinant Monoclonal
100 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide
P19256
CD58
Recombinant protein ecoding Human CD58 (Met1-Arg215).
100 Tests
Primary
Human
Antibody
IgG
Flow Cytometry
083
PerCP
CD58
Ag3; CD305; CD58; CD58 antigen; CD58 antigen, (lymphocyte function-associated antigen 3); CD58 molecule; FLJ23181; FLJ43722; LAIR1; LAIR-1; LFA3; LFA-3; Lymphocyte function-associated antigen 3; membrane receptor; RP5-1086K13.2; surface glycoprotein LFA-3
Rabbit
Protein A
RUO
965
4°C, store in dark, DO NOT FREEZE!
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado