missing translation for 'onlineSavingsMsg'
Learn More

CD59 Mouse Monoclonal Antibody, Proteintech

Product Code. p-200108204 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235531 150 μL 150µL
30237518 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235531 Supplier Proteintech Supplier No. 682221IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD59 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD59. This antibody recognizes human antigen. The CD59 antibody has been validated for the following applications: WB, IHC, IF/ICC, IF-P, ELISA analysis.

Specifications

Antigen CD59
Applications Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, ELISA
Classification Monoclonal
Clone 1E3A1
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols 16.3A5; 1F5; EJ16; EJ30; EL32; FLJ38134; FLJ92039; G344; HRF-20; HRF20; MAC-IP; MACIF; MEM43; MGC2354; MIC11; MIN1; MIN2; MIN3; MIRL; MSK21; p18-20
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 966
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 18-25 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.