missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5L Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76699AF700
This item is not returnable.
View return policy
Description
CD5L Polyclonal antibody specifically detects CD5L in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CD5L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| AIM, API6, apoptosis inhibitor 6, CD5 antigen-like, CD5 antigen-like (scavenger receptor cysteine rich family), CD5 molecule-like, CT-2, IgM-associated peptide, PRO229, Spalpha, SP-ALPHA | |
| Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of human AIM. The immunogen is located within amino acids 250 - 300 of AIM. Amino Acid Squence: CDDNWGEKEDQVV | |
| 0.1 mL | |
| Apoptosis | |
| 922 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction