missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Antibody (C6/2884R), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08281AF488
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CD6 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Human recombinant full-length CD6 protein (Uniprot: P30203) | |
| 100 μg | |
| Cytokine Research, Immunology, Signal Transduction | |
| 923 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| C6/2884R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction