missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Antibody (C6/372+3F7B5), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34585JF549
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CD6 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| Human recombinant CD6 protein (C6/372); Human rheumatoid synovial T cell line ST-1 (3F7B5) (Uniprot: P30203) | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 923 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| C6/372+3F7B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction