Learn More
Novus Biologicals™ ALCAM/CD166 Antibody (AT002), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28296-1mg
 View more versions of this product
Product Code. 30045072
  • € 920.00 / 1mg
Description
ALCAM/CD166 Monoclonal antibody specifically detects ALCAM/CD166 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ALCAM/CD166
Monoclonal
Unconjugated
Q13740
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
AT002
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733
ALCAM/ CD166
1 mg
Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
214