missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD6 Antibody (SPV-L14), PerCP , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11571PCP
This item is not returnable.
View return policy
Description
CD6 Monoclonal antibody specifically detects CD6 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| CD6 | |
| Monoclonal | |
| PerCP | |
| CD6 antigenFLJ44171, CD6 molecule, T12, Tp120, TP120T-cell differentiation antigen CD6 | |
| CD8+ cytotoxic T-cell clone | |
| 0.1 mL | |
| Cytokine Research, Immunology, Signal Transduction | |
| 923 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| SPV-L14 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction