missing translation for 'onlineSavingsMsg'
Learn More

CD61 Mouse anti-Human, PE, Clone: VIPL2, Proteintech

Product Code. 16811756 Shop All Proteintech Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 Tests
missing translation for 'unitSize'
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16811756 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16811756 missing translation for 'mfr' Proteintech missing translation for 'supplierNo' PE65185100TESTS

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD61 antibody from Proteintech is a mouse monoclonal antibody generated with n/a of human CD61. This antibody recognizes human, non-human primates antigen. The CD61 antibody has been validated for the following applications: FC analysis. This antibody is pe.

Specifications

Antigen CD61
Applications Flow Cytometry
Classification Monoclonal
Clone VIPL2
Concentration 10 μL/Test
Conjugate PE
Host Species Mouse
Quantity 100 Tests
Primary or Secondary Primary
Gene ID (Entrez) 3690
Target Species Human, Non-human Primate
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.