missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD62E/CD62P Monoclonal Antibody (1.2B6), PE
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA528649

Product Code. 16392904

  • € 314.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Beskrivning

Beskrivning

CD62E/CD62P Monoclonal Antibody for Flow

CD62E represents the 97 kDa type I transmembrane glycoprotein E-selectin, also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1). It is expressed on cytokine-stimulated endothelium, weakly on unactivated endothelium, and is absent on leukocytes. Like other selectins, CD62E is an adhesion molecule that contributes to the initial attachment and rolling of leukocytes on endothelial surfaces, a prerequisite for leukocyte extravasation into tissues.CD62P is a 140 kDa membrane glycoprotein formerly known as platelet activation-dependent granule membrane protein (PADGEM) or GMP-140. It is expressed on platelets, megakaryocytes, and epithelium. It is thought to mediate the adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses and of activated platelets to neutrophils and monocytes in hemostasis. The monoclonal antibody 1.2B6 recognizes a common epitope shared by CD62E and CD62P.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

CD62E/CD62P
Monoclonal
0.1 mg/mL
PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5
P16109, P16581, P98110
SELE, SELP
TNF stimulated human endothelial cells.
1 mL
Primary
Human, Pig
Antibody
IgG1 κ
Flow Cytometry
1.2B6
PE
SELP
CD62; CD62 antigen-like family member E; CD62 antigen-like family member P; CD62E; CD62P; CD62P; CD62 homolog; ELAM; ELAM1; ELAM-1; endothelial adhesion molecule 1; endothelial leukocyte adhesion molecule 1; ESEL; E-selectin; GMP140; GMP-140; GMRP; Granule membrane protein 140; granule membrane protein 140kDa; granulocyte membrane protein; GRMP; LECAM2; LECAM-2; LECAM3; LECAM-3; leukocyte endothelial cell adhesion molecule 2; leukocyte-endothelial cell adhesion molecule 2; Leukocyte-endothelial cell adhesion molecule 3; PADGEM; Platelet activation dependent granule-external membrane protein; platelet alpha-granule membrane protein; PSEL; P-selectin; P-selectin precursor; RP1-117P20.2; SELE; selectin E; selectin E (endothelial adhesion molecule 1); selectin P; selectin P (granule membrane protein 140kDa, antigen CD62); SELP
Mouse
Purified
RUO
397150, 397508, 6401, 6403
4°C, store in dark, DO NOT FREEZE!
Liquid
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.