missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L Armenian Hamster anti-Rat, BB700, Clone: HRL1, BD OptiBuild™
Armenian Hamster Monoclonal Antibody
Brand: BD Biosciences 745917
This item is not returnable.
View return policy
Description
CD62L Monoclonal antibody specifically detects CD62L in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD62L | |
| Monoclonal | |
| 0.2 mg/mL | |
| Armenian Hamster | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| HRL1 | |
| Brilliant Blue 700 (BB700) | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG2 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction