Learn More
Invitrogen™ CD62L (L-Selectin) Monoclonal Antibody (DREG-56 (DREG56)), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
€ 198.00 - € 360.00
Specifications
| Antigen | CD62L (L-Selectin) |
|---|---|
| Clone | DREG-56 (DREG56) |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17433372
![]() |
Invitrogen™
367-0629-42 |
100 Tests |
€ 360.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17423372
|
Invitrogen™
367-0629-41 |
25 Tests |
€ 198.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DREG-56 monoclonal antibody reacts with human CD62L, a 76 kDa member of the selectin family. CD62L is expressed by neutrophils, monocytes, and subsets of T, B, and NK cells and binds a number of glycosylated, fucosylated, sulfated sialylated glycoproteins including CD34, glycam-1 and MAdCAM-1. These interactions mediate rolling of lymphocytes on activated endothelium at the sites of inflammation and homing of cells to the high endothelial venules (HEV) of peripheral lymphoid tissues. Applications Reported: This DREG-56 (DREG56) antibody has been reported for use in flow cytometric analysis. Applications Tested: This DREG-56 (DREG56) antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Brilliant Ultraviolet 737 is a tandem dye that emits at 732 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser.
The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.Specifications
| CD62L (L-Selectin) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| P14151 | |
| 6402 | |
| IgG1 κ | |
| Affinity chromatography | |
| SELL |
| DREG-56 (DREG56) | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| Mouse | |
| Human | |
| A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1 | |
| SELL | |
| Primary | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.