missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD62L (L-Selectin) Rat anti-Mouse, BUV615, Clone: MEL-14, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 752311
This item is not returnable.
View return policy
Description
CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD62L (L-Selectin) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Ms CD62L BUV615 MEL-14 50 μg | |
| C3H/eb mouse B lymphoma 38C-13 | |
| RUO | |
| CD62L (L-Selectin) | |
| Refrigerate (2 to 8°C) | |
| Mouse |
| Flow Cytometry | |
| MEL-14 | |
| Brilliant Ultraviolet 615 | |
| Rat | |
| 1 x 50 μg | |
| Primary | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction