Learn More
CD62L (L-Selectin) Rat anti-Mouse, BUV737, Clone: MEL-14, BD Horizon™
Rat Monoclonal Antibody
Brand:  BD Biosciences 612833
Product Code. 17227924
  • ONLINE EXCLUSIVE PRICE
    € 486.00
    € 473.62 / 50µg
    Save € 12.38
    2% Off
Description
CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD62L (L-Selectin)
Monoclonal
0.2 mg/mL
Rat
RUO
CD62L (L-Selectin)
Refrigerate (2 to 8°C)
Flow Cytometry
MEL-14
Brilliant Ultraviolet 737
50 μg
Primary
Mouse
IgG2a κ