missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD62L Monoclonal Antibody (FMC46)
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 528.00 - € 794.00

Specifications

Antigen CD62L
Clone FMC46
Concentration 1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11514093
GREENER_CHOICE
View Documents
Invitrogen™
MA134177
100 μg
€ 528.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11524093
View Documents
Invitrogen™
MA134180
200 μg
€ 794.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Descrizione

Descrizione

CD62L Monoclonal Antibody for IHC (F), Flow, IP

The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Specifica

Specifica

CD62L
1 mg/mL
Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Unconjugated
Mouse
RUO
PBS with 0.09% sodium azide
P14151, P98131, Q95198
102142895, 281485, 480080, 6402, 701419
PHA stimulated lymphoblasts.
Antibody
FMC46
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Monoclonal
Liquid
IgG2b
Human, Canine, Bovine, Rhesus Macaque, Cynomolgus Monkey
SELL
A.11; AI528707; CD62 antigen-like family member L; CD62L; gp90-MEL; hLHRc; LAM1; LAM-1; LECAM1; LECAM-1; LEU8; Leu-8; Leukocyte adhesion molecule 1; leukocyte surface antigen Leu-8; Leukocyte-endothelial cell adhesion molecule 1; LNHR; LSEL; L-selectin; Ly22; Ly-22; LYAM1; Lyam-1; Ly-m22; Lymph node homing receptor; lymphocyte adhesion molecule 1; lymphocyte antigen 22; Lymphocyte surface MEL-14 antigen; pln homing receptor; PLNHR; SELE; selectin E (endothelial adhesion molecule 1); selectin L; selectin L (lymphocyte adhesion molecule 1); selectin, lymphocyte; Sell; TQ1
SELL
Primary
Protein G
Videos
SDS
Documenti

Documenti

Certificati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato