Learn More
CD63 Antibody (2585C), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB50481MFV450
Product Code. 18502546
  • € 629.00 / 0.10mL
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD63
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
0.1 mL
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
967
Store at 4C in the dark.
Flow Cytometry, Immunocytochemistry, CyTOF
2585C
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30
Chinese Hamster Ovary cell line CHO-derived human CD63, Ala63-Val203, Accession # P08962-1