Learn More
CD63 Antibody (2585J), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals FAB50482N
Product Code. 30065936
  • € 579.00 / 0.10mL
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Flow (Intracellular), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD63
Monoclonal
Alexa Fluor 700
50mM Sodium Borate
Rabbit
Protein A or G purified from cell culture supernatant
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
2585J
Western Blot, Simple Western, Immunohistochemistry, Flow (Intracellular), CyTOF-ready
CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30
Chinese Hamster Ovary cell line CHO-derived human CD63, Ala103-Val203, Accession # P08962-1
0.1 mL
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis
967
Store at 4C in the dark.