missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (2585J), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB50482H
This item is not returnable.
View return policy
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| CD63 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 2585J | |
| Western Blot, Simple Western, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Chinese Hamster Ovary cell line CHO-derived human CD63, Ala103-Val203, Accession # P08962-1 | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction