missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (7G4.2E8), Janelia Fluor™ 646, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-36567JF646
This item is not returnable.
View return policy
Description
CD63 Monoclonal specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| CD63 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD63 | |
| Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG2a λ |
| Western Blot, Flow Cytometry | |
| 7G4.2E8 | |
| Western Blot, Flow Cytometry | |
| CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin | |
| Rat | |
| Protein G purified | |
| Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction