missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (AD1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68077APC-100UL
This item is not returnable.
View return policy
Description
CD63 Monoclonal specifically detects CD63 in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD63 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% sodium azide | |
| CD63 | |
| RBL-2H3 rat basophilic leukemia cells. | |
| Primary | |
| Rat | |
| IgG1 |
| Flow Cytometry | |
| AD1 | |
| Flow Cytometry | |
| CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin | |
| Mouse | |
| 100 μL | |
| 967 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction