missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD63 Antibody (H5C6), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42225AF647
This item is not returnable.
View return policy
Description
CD63 Monoclonal antibody specifically detects CD63 in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay, In vitro Assay, Dot Blot, Electron Microscopy, Flow (Intracellular), Immunohistochemistry (Whole-Mount), Neutralization
Specifications
| CD63 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| CD_antigen: CD63, CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, lysosome-associated membrane glycoprotein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30 | |
| Human splenic adherent cells. | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis | |
| 967 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| H5C6 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction