Learn More
CD63 Antibody (LAMP3/968), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-47936MFV610
Product Code. 18669559
  • € 827.00 / 0.10mL
Description
CD63 Monoclonal antibody specifically detects CD63 in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flow Cytometry
LAMP3/968
Flow Cytometry, Flow (Intracellular)
Primary
Monoclonal
mFluor Violet 610 SE
Autophagy, Cancer, Cardiovascular Biology, Cellular Signaling, Cytokine Research, Immunology, Innate Immunity, Lysosome Markers, Metastasis