missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CBR1 Mouse anti-Human, Clone: OTI4B4, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809840S
This item is not returnable.
View return policy
Description
CBR1 Monoclonal antibody specifically detects CBR1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CBR1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16152 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 873 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B4 | |
| Unconjugated | |
| CBR1 | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NADP(+)]; 20-beta-hydroxysteroid dehydrogenase; AW261796; carbonyl reductase (NADPH) 1; carbonyl reductase [NADPH] 1; Carbonyl reductase [NADPH] 1 (NADPH-dependent carbonyl reductase 1); carbonyl reductase 1; CBR; Cbr 1; Cbr1; CR; CRN; hCBR1; NADPH-dependent carbonyl reductase 1; prostaglandin 9-ketoreductase; Prostaglandin-E(2) 9-reductase; SDR21C1; Short chain dehydrogenase/reductase family 21C member 1; short chain dehydrogenase/reductase family 21C, member 1 | |
| Full length human recombinant protein of human CBR1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction