missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERCC3 Mouse anti-Human, Clone: OTI6H8, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809371
This item is not returnable.
View return policy
Description
The human XPB DNA helicase is a subunit of the DNA repair/basal transcription factor TFIIH, and is involved in early steps of the nucleotide excision repair (NER) pathway. Two distinct clinical phenotypes, xeroderma pigmentosum associated with Cockayne′s syndrome (XP/CS) and trichothiodystrophy (TTD), can be due to mutations in the XPB gene.Specifications
| ERCC3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P19447 | |
| ERCC3 | |
| Human recombit protein fragment corresponding to amino acids 191-436 of human ERCC3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6H8 | |
| Unconjugated | |
| ERCC3 | |
| BTF2, GTF2H, RAD25, TFIIH, TTD2, XPB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2071 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction