Learn More
CD68 Monoclonal Antibody (OTI6C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA803046S
Encompass_Preferred
 View more versions of this product
Product Code. 17591933
  • € 242.00 / 30µL
Description
CD68 Monoclonal antibody specifically detects CD68 in Human samples. It is validated for Western Blot
Specifications
Specifications
CD68
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q13506
Mouse
Affinity chromatography
RUO
4664
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI6C3
Unconjugated
NAB1
CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen
Human recombinant protein fragment corresponding to amino acids 237-487 of human NAB1 produced in E.coli.