missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68 Monoclonal Antibody (OTI6D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802966S
This item is not returnable.
View return policy
Description
CD68 Monoclonal antibody specifically detects CD68 in Human samples. It is validated for Western BlotSpecifications
| CD68 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P34810 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 968 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI6D3 | |
| Unconjugated | |
| Cd68 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Human recombinant protein fragment corresponding to amino acids 22-319 of human CD68 produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction