missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR92 Monoclonal Antibody (OTI4B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809523S
This item is not returnable.
View return policy
Description
WDR92 Monoclonal antibody specifically detects WDR92 in Human samples. It is validated for Western BlotSpecifications
| WDR92 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96MX6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 116143 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI4B9 | |
| Unconjugated | |
| WDR92 | |
| AI553587; HZGJ; monad; RGD1563863; testicular secretory protein Li 67; WD repeat domain 92; WD repeat-containing protein 92; WD repeat-containing protein Monad; Wdr92 | |
| Full length human recombinant protein of human WDR92 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction