Learn More
CD68 Monoclonal Antibody (OTI13H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802981
Encompass_Preferred
 View more versions of this product
Product Code. 17531853
  • € 540.87 / 100µL
Description
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predomitly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human maligcies, including several Acute Myeloid Leukemia studies.
Specifications
Specifications
CD68
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P34810
CD68
Human recombit protein fragment corresponding to amino acids 22-319 of human CD68 produced in SF9 cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI13H2
Unconjugated
CD68
GP110, LAMP4, SCARD1
Mouse
Affinity Chromatography
RUO
968
-20° C, Avoid Freeze/Thaw Cycles