Learn More
CD68 Mouse, anti-Human, CoraLite Plus 405, Clone: Y1/82A, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL405-65187-100TESTS
Description
The CD68 antibody from Proteintech is a mouse monoclonal antibody generated with lysosomal contents of lung macrophages of human CD68. This antibody recognizes human antigen. The CD68 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 405.
Specifications
| CD68 | |
| Y1/82A | |
| CoraLite Plus 405 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2b κ |
| Flow Cytometry (Intracellular Staining) | |
| 5 μL/Test | |
| CD68, CD68 molecule, DKFZp686M18236, GP110, Macrosialin, SCARD1 | |
| 100 Tests | |
| 968 | |
| Store at 2°C to 8°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.