missing translation for 'onlineSavingsMsg'
Learn More

CD68 Mouse, anti-Human, CoraLite Plus 750, Clone: 3A9A7, Proteintech

Product Code. 18074705 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074705 20 μL 20µL
18048075 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074705 Supplier Proteintech Supplier No. CL7506623120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The CD68 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD68. This antibody recognizes human, mouse antigen. The CD68 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 750.

Specifications

Antigen CD68
Applications Western Blot
Clone 3A9A7
Concentration 1000 μg/mL
Conjugate CoraLite Plus 750
Gene Symbols CD68, CD68 molecule, DKFZp686M18236, GP110, Macrosialin, SCARD1
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 968
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 155 kDa
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.