missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD68 Recombinant Mouse Monoclonal Antibody (KP-1)

Mouse Recombinant Monoclonal Antibody
€ 156.00 - € 474.00
Specifications
| Antigen | CD68 |
|---|---|
| Clone | KP-1 |
| Concentration | 50 μg/mL |
| Content And Storage | 4°C |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17715569
![]() |
Invitrogen™
MA544195 |
100 μL |
€ 474.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17725569
|
Invitrogen™
MA544196 |
10 μL |
€ 156.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD68 Recombinant Monoclonal Antibody for IHC (P), Flow
CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.Specifications
| CD68 | |
| 50 μg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 0.1% BSA and 0.09% sodium azide; pH 6.8-7.4 | |
| P34810 | |
| 968 | |
| Lysosomal Fraction Of Human Lung Macrophages. | |
| Antibody |
| KP-1 | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG1 | |
| Human | |
| Cd68 | |
| CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen | |
| Cd68 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.