missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (001), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90914H
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| This CD68/SR-D1 recombinant Antibody (001) was developed against purified, recombinant rat CD68/SR-D1 (Q4FZY1; Met1-Ser295). | |
| 0.1 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction