missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (2449C), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10114G
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 2449C | |
| Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Chinese Hamster Ovary cell line, CHO-derived mouse CD68, Asp21-Pro282, Accession # P31996 | |
| 0.1 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction