missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (FA-11), DyLight 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-33337UV
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| DyLight 350 | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| FA-11 | |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| CD68 | |
| This CD68/SR-D1 Antibody (FA-11) was developed against purified ConA acceptor glycoproteins from the P815 cell line. | |
| 0.1 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction