missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (OTI1E6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70381FR
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blot.
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| CD68 | |
| This CD68/SR-D1 Antibody (OTI1E6) was developed against human recombinant protein fragment corresponding to amino acids 22-319 of human CD68 (NP_001242)produced in SF9 cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Western Blot | |
| OTI1E6 | |
| Western Blot | |
| CD68 antigenmacrophage antigen CD68, CD68 molecule, DKFZp686M18236, GP110, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| Mouse | |
| Affinity Purified | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction