Learn More
CD68/SR-D1 Antibody (SPM130), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-34736MFV500
Product Code. 18755060
  • € 782.00 / 0.10mL
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human, Feline, Monkey, Rabbit, Canine (Negative), Chicken (Negative), Porcine (Negative) samples. It is validated for Flow Cytometry