missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD68/SR-D1 Antibody (Y-2/131/47), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50149JF646
This item is not returnable.
View return policy
Description
CD68/SR-D1 Monoclonal antibody specifically detects CD68/SR-D1 in Human samples. It is validated for Western Blot
Specifications
| CD68/SR-D1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Y-2/131/47 | |
| Western Blot | |
| CD_antigen: CD68, CD68, CD68 antigen, CD68 molecule, DKFZp686M18236, GP110, LAMP4, macrophage antigen CD68, macrosialin, SCARD1, scavenger receptor class D, member 1 | |
| This CD68/SR-D1 Antibody (Y-2/131/47) was developed against Phytohaemagglutinin-activated peripheral blood mononuclear cells | |
| 0.1 mL | |
| Cell Biology, Cytokine Research, Immunology, Microglia Markers | |
| 968 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction