missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (15B5G2) - Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 548.00
Specifications
| Antigen | CD69 |
|---|---|
| Clone | 15B5G2 |
| Concentration | 0.5mg/mL |
| Dilution | Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1-3 ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18183315
|
Novus Biologicals
NBP2-25242SS |
0.025 mg |
€ 220.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18006664
|
Novus Biologicals
NBP2-25242 |
0.1 mg |
€ 548.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD69 Monoclonal specifically detects CD69 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD69 | |
| 0.5mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| CD69 | |
| IgG2a κ | |
| Protein G purified | |
| This product reacts with Human. |
| 15B5G2 | |
| Western Blot 1-5 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1-3 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| NP_001772 | |
| 969 | |
| Amino acids 103-199 of human CD69 were used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title