missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD69 Antibody (FN50) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-43387JF525
This item is not returnable.
View return policy
Description
CD69 Monoclonal antibody specifically detects CD69 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen)
Specifications
| CD69 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Activation inducer molecule, activation inducer molecule (AIM/CD69), AIM, BL-AC/P26, CD69 antigen, CD69 antigen (p60, early T-cell activation antigen), CD69 molecule, CLEC2CC-type lectin domain family 2 member C, C-type lectin domain family 2, member C, EA1, early activation antigen CD69, early lymphocyte activation antigen, Early T-cell activation antigen p60, GP32/28, Leukocyte surface antigen Leu-23, MLR-3 | |
| The immunogen for this antibody was CD69. | |
| 0.1 mL | |
| Innate Immunity | |
| 969 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| FN50 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction