missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Antibody (CD7/8357R), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20950IR
This item is not returnable.
View return policy
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD7 | |
| Monoclonal | |
| DyLight 755 | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| Recombinant fragment (around aa 1-200) of human CD7 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CD7/8357R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction