missing translation for 'onlineSavingsMsg'
Learn More

CD7 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-32097R

Product Code. 18404336

  • € 634.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD7 Polyclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD7
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
The immunogen this antibody was made to, maps to a region between residue 190 to 240 of human CD7 Molecule using the numbering given in entry NP_006128.1 (GeneID 924).
0.1 mL
Cytokine Research, Signal Transduction
924
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 550
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.