missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Antibody (MEM-186), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-326UV
This item is not returnable.
View return policy
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
Specifications
| CD7 | |
| Monoclonal | |
| DyLight 350 | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| Human acute myelogenous leukaemia cell line KG-1. | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| MEM-186 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction