missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CD7 Antibody (Persongen patent anti-CD7), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28213-100ug
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| CD7 | |
| Monoclonal | |
| Unconjugated | |
| P09564 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Persongen patent anti-CD7 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| CD7 | |
| 100 μg | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion