missing translation for 'onlineSavingsMsg'
Learn More

CD7 Antibody (rCD7/6972), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20944AF532

619.65 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 18576458

  • € 728.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD7
Monoclonal
Alexa Fluor 532
CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein
Recombinant fragment (around aa140-240) of human CD7 protein (exact sequence is proprietary)
0.1 mL
Cytokine Research, Signal Transduction
924
Store at 4°C in the dark.
IgG1 κ
Immunohistochemistry (Paraffin)
rCD7/6972
50 mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.