missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSC22D2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5148315

Product Code. 30019895

  • € 368.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TSC22D2 Polyclonal Antibody for IHC (P)

TSC22D2 belongs to the TSC-22/Dip/Bun family and the function remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TSC22D2
Polyclonal
Unconjugated
TSC22D2
1810043J12Rik; 5530402M19Rik; cb869; id:ibd5097; kiaa0669; KIAA0669-like protein; TILZ4; TILZ4a; TILZ4b; TILZ4c; TSC22 domain family 2; TSC22 domain family member 2; TSC22 domain family protein 2; TSC22 domain family protein 2-like protein; TSC22 domain family, member 2; TSC22D2; tsc22d2 protein; TSC22-related-inducible leucine zipper protein 4; wu:fa55e04; zgc:92475
Rabbit
Antigen affinity chromatography, Protein A
RUO
9819
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
100 μg/mL
PBS with 0.03% ProClin 300; pH 7
O75157
TSC22D2
E. coli-derived Human TSC22D2 fragment.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.