missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD72 Antibody (108), Alexa Fluor™ 405 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-06096AF405
This item is not returnable.
View return policy
Description
CD72 Monoclonal antibody specifically detects CD72 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD72 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CD72 antigenB-cell differentiation antigen CD72, CD72 molecule, CD72b, Lyb-2, LYB2lyb-2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD72.1 (NP_001104530.1; Met1-Lys95). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 971 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry | |
| 108 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction